siRNA Details
| Gene Symbol: AHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029882.3 | NP_001025053.1 | 6 | 3809 | ||
| XM_005245848.3 | XP_005245905.1 | 7 | 4044 | |||
| XM_005245849.3 | XP_005245906.1 | 7 | 4149 | |||
| XM_005245850.3 | XP_005245907.1 | 8 | 4169 | |||
| XM_005245851.3 | XP_005245908.1 | 7 | 4041 | |||
| XM_005245852.3 | XP_005245909.1 | 6 | 3818 | |||
| XM_011541255.1 | XP_011539557.1 | 7 | 3723 | |||
| XM_011541256.2 | XP_011539558.1 | 7 | 4163 | |||
| XM_011541257.2 | XP_011539559.1 | 6 | 4918 | |||
| GenBank mRNA | AK125431.1 | - | 1 | 527 | ||

AHDC1
Genomic Map