siRNA Details
| Gene Symbol: SULT1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014465.3 | NP_055280.2 | 7 | 1010 | ||
| XM_005265676.3 | XP_005265733.1 | 8 | 925 | |||
| XM_005265677.3 | XP_005265734.1 | 8 | 1125 | |||
| XM_005265678.3 | XP_005265735.1 | 7 | 878 | |||
| XM_011531873.2 | XP_011530175.1 | 6 | 749 | |||
| GenBank mRNA | BC010895.1 | - | 7 | 1012 | 4105139 | |
| D89479.1 | - | 6 | 736 | |||
| U95726.1 | - | 6 | 727 | |||

SULT1B1
Genomic Map