siRNA Details
| Gene Symbol: TOR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085347.2 | NP_001078816.1 | 2 | 372 | ||
| NM_001134430.2 | NP_001127902.1 | 2 | 372 | |||
| NM_130459.3 | NP_569726.2 | 2 | 372 | |||
| XM_011518554.1 | XP_011516856.1 | 2 | 410 | |||
| GenBank mRNA | AK027677.1 | - | 2 | 307 | ||
| AK075520.1 | - | 2 | 315 | |||
| AK092388.1 | - | 1 | 885 | |||
| AY358180.1 | - | 2 | 339 | |||
| CN263138.1 | - | 2 | 374 | |||
| DA430836.1 | - | 2 | 348 | |||

TOR2A
Genomic Map