siRNA Details
| Gene Symbol: GOLGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002077.3 | NP_002068.1 | 19 | 2190 | ||
| XM_005251929.3 | XP_005251986.1 | 19 | 2122 | |||
| XM_006717062.3 | XP_006717125.1 | 19 | 2186 | |||
| XM_006717063.3 | XP_006717126.1 | 19 | 2157 | |||
| GenBank mRNA | AB208858.1 | - | 14 | 1543 | ||
| AK302445.1 | - | 19 | 1999 | |||
| BC032853.1 | - | 19 | 2160 | 4828793 | ||
| U51587.1 | - | 19 | 2182 | |||

GOLGA1
Genomic Map