siRNA Details
| Gene Symbol: GOLGA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172557.1 | NP_001166028.1 | 3 | 515 | ||
| NM_005895.3 | NP_005886.2 | 3 | 853 | |||
| XM_006719736.3 | XP_006719799.1 | 3 | 580 | |||
| XM_006719737.3 | XP_006719800.1 | 3 | 590 | |||
| GenBank mRNA | AF485338.1 | - | 2 | 297 | ||
| BC060826.1 | - | 3 | 539 | |||
| BC142658.1 | - | 3 | 515 | 40148073 | ||
| BC146675.1 | - | 3 | 515 | 40148852 | ||
| D63997.1 | - | 3 | 563 | |||

GOLGA3
Genomic Map