siRNA Details
| Gene Symbol: SFN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006142.3 | NP_006133.1 | 1 | 504 | ||
| GenBank mRNA | AF029082.1 | - | 1 | 504 | ||
| BC000329.2 | - | 1 | 484 | 2822397 | ||
| BC000995.2 | - | 1 | 481 | 3445915 | ||
| BC001550.1 | - | 2 | 390 | 3457665 | ||
| BC002995.1 | - | 1 | 484 | 2822397 | ||
| BC023552.2 | - | 1 | 508 | 3534328 | ||
| CR541905.1 | - | 1 | 433 | |||
| CR541926.1 | - | 1 | 433 | |||
| M93010.1 | - | 1 | 443 | |||

SFN
Genomic Map