siRNA Details
Gene Symbol: OR4C13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001955.2 | NP_001001955.2 | 1 | 457 | ||
GenBank mRNA | BC137005.1 | - | 1 | 457 | 9020998 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR4C13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001955.2 | NP_001001955.2 | 1 | 457 | ||
GenBank mRNA | BC137005.1 | - | 1 | 457 | 9020998 |