siRNA Details
| Gene Symbol: KLHL35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039548.2 | NP_001034637.2 | 5 | 1460 | ||
| XM_011544954.2 | XP_011543256.1 | 6 | 2969 | |||
| GenBank mRNA | AK091109.1 | - | 4 | 1855 | ||
| AL050370.1 | - | 1 | 1103 | |||
| BC042952.2 | - | 5 | 1001 | 5275230 | ||
| BC057763.1 | - | 5 | 1022 | |||
| BC131539.1 | - | 2 | 441 | |||
| BC132708.1 | - | 5 | 824 | 40146730 | ||
| BC132710.1 | - | 5 | 824 | 40146732 | ||

KLHL35
Genomic Map