siRNA Details
| Gene Symbol: ZNF740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004304.3 | NP_001004304.1 | 4 | 611 | ||
| XM_006719344.2 | XP_006719407.1 | 4 | 586 | |||
| XM_011538194.2 | XP_011536496.1 | 4 | 811 | |||
| XM_017019181.1 | XP_016874670.1 | 4 | 845 | |||
| GenBank mRNA | AK292744.1 | - | 4 | 662 | ||
| BC053557.1 | - | 4 | 614 | 6162269 | ||

ZNF740
Genomic Map