siRNA Details
| Gene Symbol: C12orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031748.3 | NP_001026918.2 | 6 | 652 | ||
| NM_001319247.1 | NP_001306176.1 | 6 | 652 | |||
| NR_135051.1 | 5 | 513 | ||||
| XM_005268806.3 | XP_005268863.1 | 5 | 462 | |||
| XM_011538231.2 | XP_011536533.1 | 5 | 689 | |||
| GenBank mRNA | AK097445.1 | - | 5 | 462 | ||
| AL705293.1 | - | 6 | 628 | |||
| BC038754.1 | - | 6 | 649 | 5269851 | ||
| BC048120.1 | - | 6 | 636 | 5266211 | ||
| BI461967.1 | - | 6 | 653 | |||
| EU668334.1 | - | 5 | 462 | |||
| GQ472216.1 | - | 5 | 462 | |||

C12orf40
Genomic Map