siRNA Details
| Gene Symbol: SIAH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198849.2 | NP_942146.2 | 1 | 118 | ||
| GenBank mRNA | BC041372.2 | - | 1 | 118 | ||
| DA804734.1 | - | 1 | 143 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SIAH3| Gene Symbol: SIAH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198849.2 | NP_942146.2 | 1 | 118 | ||
| GenBank mRNA | BC041372.2 | - | 1 | 118 | ||
| DA804734.1 | - | 1 | 143 | |||