siRNA Details
| Gene Symbol: REC114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042367.1 | NP_001035826.1 | 1 | 117 | ||
| GenBank mRNA | AK131030.1 | - | 1 | 111 | ||
| BC055412.1 | - | 1 | 98 | |||
| DB465854.1 | - | 1 | 117 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

REC114| Gene Symbol: REC114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042367.1 | NP_001035826.1 | 1 | 117 | ||
| GenBank mRNA | AK131030.1 | - | 1 | 111 | ||
| BC055412.1 | - | 1 | 98 | |||
| DB465854.1 | - | 1 | 117 | |||