siRNA Details
| Gene Symbol: TMEM114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146336.1 | NP_001139808.1 | 4 | 458 | ||
| NM_001290095.1 | NP_001277024.1 | 3 | 320 | |||
| NM_001290097.1 | NP_001277026.1 | 4 | 412 | |||
| GenBank mRNA | BC150511.1 | - | 2 | 235 | ||
| BE670838.1 | - | 3 | 252 | |||
| BM670302.1 | - | 1 | 499 | |||
| BM712244.1 | - | 4 | 412 | |||

TMEM114
Genomic Map