siRNA Details
| Gene Symbol: TSEN54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207346.2 | NP_997229.2 | 6 | 531 | ||
| XM_005257229.3 | XP_005257286.1 | 6 | 530 | |||
| GenBank mRNA | AK094466.1 | - | 6 | 515 | ||
| AK097583.1 | - | 6 | 1122 | |||
| BC045551.1 | - | 6 | 505 | |||
| BC047793.1 | - | 6 | 500 | 6500549 | ||
| BC053643.1 | - | 6 | 502 | |||
| BX424294.2 | - | 6 | 511 | |||

TSEN54
Genomic Map