siRNA Details
| Gene Symbol: HEXDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173620.2 | NP_775891.2 | 3 | 549 | ||
| XM_005256347.4 | XP_005256404.1 | 3 | 273 | |||
| XM_006722280.3 | XP_006722343.1 | 2 | 112 | |||
| XM_011523561.2 | XP_011521863.1 | 1 | 20 | |||
| XM_017024484.1 | XP_016879973.1 | 3 | 273 | |||
| GenBank mRNA | AK074405.1 | - | 2 | 112 | ||
| BC018205.1 | - | 2 | 144 | 3464710 | ||
| BC035561.1 | - | 3 | 230 | 5211583 | ||
| DN999062.1 | - | 3 | 555 | |||
| FM204887.1 | - | 2 | 133 | |||

HEXDC
Genomic Map