siRNA Details
| Gene Symbol: PTRF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012232.5 | NP_036364.2 | 2 | 874 | ||
| GenBank mRNA | AF000421.1 | - | 1 | 93 | ||
| AK000715.1 | - | 2 | 360 | |||
| AK092406.1 | - | 3 | 675 | |||
| AK298072.1 | - | 3 | 755 | |||
| AK298563.1 | - | 3 | 729 | |||
| AK314389.1 | - | 2 | 810 | |||
| BC004295.1 | - | 1 | 84 | |||
| BC008849.1 | - | 2 | 157 | 4140111 | ||
| BC066123.1 | - | 2 | 810 | 5269172 | ||
| BC073759.1 | - | 2 | 696 | 6572478 | ||

PTRF
Genomic Map