siRNA Details
Gene Symbol: LOC101928710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110848.1 | 4 | 503 | |||
NR_110849.1 | 4 | 612 | ||||
GenBank mRNA | BG613706.1 | - | 4 | 613 | ||
BX362577.2 | - | 4 | 497 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110848.1 | 4 | 503 | |||
NR_110849.1 | 4 | 612 | ||||
GenBank mRNA | BG613706.1 | - | 4 | 613 | ||
BX362577.2 | - | 4 | 497 |