siRNA Details
| Gene Symbol: SSC5D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144950.1 | NP_001138422.1 | 13 | 2959 | ||
| GenBank mRNA | EU699476.1 | - | 13 | 2895 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SSC5D| Gene Symbol: SSC5D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144950.1 | NP_001138422.1 | 13 | 2959 | ||
| GenBank mRNA | EU699476.1 | - | 13 | 2895 | ||