siRNA Details
| Gene Symbol: CXCL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198477.2 | NP_940879.1 | 4 | 561 | ||
| NR_133910.1 | 4 | 578 | ||||
| GenBank mRNA | AK292985.1 | - | 4 | 459 | ||
| AY358433.1 | - | 4 | 497 | |||
| AY598464.1 | - | 4 | 561 | |||
| BC093946.1 | - | 4 | 412 | 7939791 | ||
| BC112095.1 | - | 4 | 414 | 8327563 | ||
| BM978124.1 | - | 1 | 370 | |||
| BU684368.1 | - | 1 | 374 | |||

CXCL17
Genomic Map