siRNA Details
| Gene Symbol: NKPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198478.3 | NP_940880.3 | 4 | 762 | ||
| XM_011526799.2 | XP_011525101.1 | 5 | 3001 | |||
| XM_011526802.2 | XP_011525104.1 | 5 | 2740 | |||
| XM_011526804.2 | XP_011525106.1 | 4 | 2563 | |||
| XM_011526805.2 | XP_011525107.1 | 3 | 619 | |||
| GenBank mRNA | BC117228.1 | - | 1 | 178 | 40125779 | |
| BC143792.1 | - | 1 | 178 | 9052306 | ||
| EG328190.1 | - | 3 | 273 | |||
| EG328439.1 | - | 1 | 61 | |||

NKPD1
Genomic Map