siRNA Details
| Gene Symbol: GPR21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005294.2 | NP_005285.1 | 2 | 1262 | ||
| XM_005251933.3 | XP_005251990.1 | 2 | 7469 | |||
| GenBank mRNA | AK313539.1 | - | 2 | 1236 | ||
| BC066885.1 | - | 1 | 766 | 6971731 | ||
| BC066886.1 | - | 1 | 766 | 6971734 | ||
| BC067532.1 | - | 1 | 766 | 6971732 | ||
| BC067533.1 | - | 1 | 766 | 6971733 | ||
| BC067534.1 | - | 1 | 766 | 6971735 | ||
More Information
Set Membership: |
|

GPR21
Genomic Map