siRNA Details
| Gene Symbol: OR2L13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304535.1 | NP_001291464.1 | 2 | 854 | ||
| NM_175911.3 | NP_787107.1 | 3 | 979 | |||
| XM_011544169.2 | XP_011542471.1 | 3 | 723 | |||
| GenBank mRNA | BC028158.1 | - | 3 | 817 | 5240594 | |

OR2L13
Genomic Map