siRNA Details
| Gene Symbol: RUSC1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039517.1 | NP_001034606.1 | 2 | 555 | ||
| GenBank mRNA | AK093295.1 | - | 1 | 336 | ||
| BC062571.1 | - | 2 | 584 | 5183308 | ||
| BC080538.1 | - | 2 | 555 | 5018492 | ||
| BC131614.1 | - | 1 | 317 | 40115930 | ||

RUSC1-AS1
Genomic Map