siRNA Details
| Gene Symbol: EPHA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099439.1 | NP_001092909.1 | 3 | 389 | ||
| NM_173641.2 | NP_775912.2 | 3 | 389 | |||
| GenBank mRNA | AJ781169.1 | - | 3 | 389 | ||
| AK090974.1 | - | 3 | 370 | |||
| BC067734.1 | - | 3 | 389 | 4375803 | ||

EPHA10
Genomic Map