siRNA Details
| Gene Symbol: C1orf111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182581.3 | NP_872387.2 | 3 | 546 | ||
| XM_005245103.3 | XP_005245160.1 | 2 | 540 | |||
| XM_011509443.2 | XP_011507745.1 | 3 | 711 | |||
| XM_011509444.1 | XP_011507746.1 | 3 | 558 | |||
| GenBank mRNA | AY248900.1 | - | 3 | 525 | ||
| BC032957.1 | - | 3 | 553 | 4827863 | ||

C1orf111
Genomic Map