siRNA Details
| Gene Symbol: RIMKLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173642.3 | NP_775913.2 | 5 | 1145 | ||
| XM_006710585.3 | XP_006710648.1 | 5 | 1023 | |||
| XM_017001082.1 | XP_016856571.1 | 6 | 1204 | |||
| GenBank mRNA | BC039737.1 | - | 5 | 1139 | 6021672 | |

RIMKLA
Genomic Map