siRNA Details
| Gene Symbol: LOC285095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135766.1 | 3 | 370 | |||
| NR_135767.1 | 3 | 330 | ||||
| NR_135768.1 | 3 | 269 | ||||
| GenBank mRNA | AA193593.1 | - | 1 | 259 | ||
| AA250751.1 | - | 3 | 297 | |||
| BX108328.1 | - | 3 | 336 | |||
| DB296165.1 | - | 1 | 254 | |||
| HY129633.1 | - | 3 | 371 | |||

LOC285095
Genomic Map