siRNA Details
| Gene Symbol: ERICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290030.1 | NP_001276959.1 | 6 | 842 | ||
| XM_011510998.2 | XP_011509300.1 | 8 | 974 | |||
| XM_011510999.2 | XP_011509301.1 | 6 | 545 | |||
| XM_011511001.2 | XP_011509303.1 | 6 | 1320 | |||
| XM_017003875.1 | XP_016859364.1 | 7 | 1343 | |||
| XM_017003876.1 | XP_016859365.1 | 6 | 358 | |||
| XM_017003877.1 | XP_016859366.1 | 7 | 641 | |||

ERICH2
Genomic Map