siRNA Details
| Gene Symbol: ZNF621 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098414.2 | NP_001091884.1 | 5 | 1462 | ||
| NM_198484.4 | NP_940886.1 | 5 | 1698 | |||
| XM_005265079.3 | XP_005265136.1 | 5 | 1525 | |||
| XM_005265080.3 | XP_005265137.1 | 4 | 1444 | |||
| XM_006713114.3 | XP_006713177.1 | 5 | 1406 | |||
| XM_011533610.2 | XP_011531912.1 | 5 | 2032 | |||
| XM_017006226.1 | XP_016861715.1 | 4 | 1325 | |||
| GenBank mRNA | AK127181.1 | - | 5 | 1462 | ||
| BC101619.1 | - | 5 | 1336 | 8069125 | ||
| BC113415.1 | - | 5 | 1336 | 8322467 | ||
| CR749268.1 | - | 5 | 1563 | |||

ZNF621
Genomic Map