siRNA Details
| Gene Symbol: ZNF852 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287349.1 | NP_001274278.1 | 4 | 330 | ||
| GenBank mRNA | AK131507.1 | - | 1 | 2499 | ||
| AK296954.1 | - | 4 | 326 | |||
| BC014381.1 | - | 3 | 219 | |||
| DC398999.1 | - | 4 | 331 | |||

ZNF852
Genomic Map