siRNA Details
| Gene Symbol: FRYL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015030.1 | NP_055845.1 | 39 | 5261 | ||
| XM_005248090.4 | XP_005248147.1 | 40 | 5319 | |||
| XM_005248093.4 | XP_005248150.1 | 14 | 1946 | |||
| XM_011513680.2 | XP_011511982.1 | 37 | 4781 | |||
| XM_011513681.2 | XP_011511983.1 | 37 | 4781 | |||
| XM_011513682.2 | XP_011511984.1 | 36 | 4736 | |||
| XM_011513683.2 | XP_011511985.1 | 39 | 5155 | |||
| XM_011513684.2 | XP_011511986.1 | 14 | 1952 | |||
| XM_017008046.1 | XP_016863535.1 | 40 | 5586 | |||
| GenBank mRNA | AL833170.1 | - | 6 | 782 | ||

FRYL
Genomic Map