siRNA Details
| Gene Symbol: RGMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012761.2 | NP_001012779.2 | 4 | 828 | ||
| XM_005271966.3 | XP_005272023.1 | 4 | 684 | |||
| XM_011543345.2 | XP_011541647.1 | 4 | 589 | |||
| XM_011543346.2 | XP_011541648.1 | 2 | 966 | |||
| XM_011543347.2 | XP_011541649.2 | 3 | 512 | |||
| XM_017009390.1 | XP_016864879.1 | 4 | 1076 | |||
| XM_017009391.1 | XP_016864880.1 | 4 | 974 | |||
| GenBank mRNA | AK074887.1 | - | 4 | 1075 | ||
| BC067736.1 | - | 4 | 828 | 30340500 | ||
| BG251558.1 | - | 2 | 406 | |||

RGMB
Genomic Map