siRNA Details
Gene Symbol: LINC00632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028344.1 | 3 | 130 | |||
GenBank mRNA | AK054978.1 | - | 3 | 122 | ||
DA082671.1 | - | 3 | 130 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028344.1 | 3 | 130 | |||
GenBank mRNA | AK054978.1 | - | 3 | 122 | ||
DA082671.1 | - | 3 | 130 |