siRNA Details
| Gene Symbol: H2BFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164416.1 | NP_001157888.1 | 1 | 121 | ||
| XM_006724640.2 | XP_006724703.1 | 1 | 154 | |||
| XM_011530922.2 | XP_011529224.1 | 1 | 317 | |||
| GenBank mRNA | AK093522.1 | - | 1 | 121 | ||

H2BFM
Genomic Map