siRNA Details
Gene Symbol: SPCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014041.3 | NP_054760.3 | 2 | 476 | ||
GenBank mRNA | AF092138.1 | - | 2 | 205 | ||
AK027426.1 | - | 2 | 205 | |||
AX775783.1 | - | 2 | 205 | |||
AX775785.1 | - | 3 | 990 | |||
AX775787.1 | - | 2 | 179 | |||
AX775833.1 | - | 2 | 195 | |||
BC000884.1 | - | 2 | 179 | 3462307 | ||
BI826431.1 | - | 2 | 170 | |||
BQ423345.1 | - | 2 | 212 | |||
BU070128.1 | - | 2 | 413 |
