siRNA Details
| Gene Symbol: C19orf53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014047.2 | NP_054766.1 | 1 | 57 | ||
| GenBank mRNA | AF078852.1 | - | 1 | 31 | ||
| AK311855.1 | - | 1 | 22 | |||
| BC015465.2 | - | 1 | 32 | 3914049 | ||
| DA768829.1 | - | 1 | 58 | |||

C19orf53
Genomic Map