siRNA Details
| Gene Symbol: FLVCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014053.3 | NP_054772.1 | 3 | 1120 | ||
| XM_011509446.2 | XP_011507748.1 | 3 | 1096 | |||
| XM_011509447.2 | XP_011507749.1 | 5 | 1338 | |||
| XM_011509448.1 | XP_011507750.1 | 5 | 1354 | |||
| GenBank mRNA | AF118637.1 | - | 3 | 1120 | ||
| AK001419.1 | - | 3 | 297 | |||
| AK314481.1 | - | 3 | 1096 | |||
| BC048312.1 | - | 3 | 1058 | 4417876 | ||

FLVCR1
Genomic Map