siRNA Details
| Gene Symbol: FLVCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014053.3 | NP_054772.1 | 5 | 1372 | ||
| XM_011509446.2 | XP_011507748.1 | 5 | 1348 | |||
| GenBank mRNA | AF118637.1 | - | 5 | 1372 | ||
| AK001419.1 | - | 5 | 549 | |||
| AK314481.1 | - | 5 | 1348 | |||
| BC048312.1 | - | 5 | 1310 | 4417876 | ||

FLVCR1
Genomic Map