siRNA Details
| Gene Symbol: C16orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014117.2 | NP_054836.2 | 3 | 779 | ||
| XM_011522462.2 | XP_011520764.1 | 3 | 1130 | |||
| GenBank mRNA | AK123266.1 | - | 1 | 43 | ||
| BC113570.1 | - | 3 | 679 | 8322622 | ||
| CB959956.1 | - | 3 | 599 | |||

C16orf72
Genomic Map