siRNA Details
| Gene Symbol: KIAA0125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026800.2 | 3 | 1012 | |||
| GenBank mRNA | AK125627.1 | - | 3 | 1013 | ||
| BC016766.1 | - | 3 | 334 | 4077490 | ||
| BC030286.1 | - | 3 | 346 | 5223261 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA0125| Gene Symbol: KIAA0125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026800.2 | 3 | 1012 | |||
| GenBank mRNA | AK125627.1 | - | 3 | 1013 | ||
| BC016766.1 | - | 3 | 334 | 4077490 | ||
| BC030286.1 | - | 3 | 346 | 5223261 | ||