siRNA Details
| Gene Symbol: KIAA0125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026800.2 | 3 | 1056 | |||
| GenBank mRNA | AF150199.1 | - | 3 | 366 | ||
| AF161538.1 | - | 3 | 366 | |||
| AK125627.1 | - | 3 | 1057 | |||
| BC016766.1 | - | 3 | 378 | 4077490 | ||
| BC030286.1 | - | 3 | 390 | 5223261 | ||

KIAA0125
Genomic Map