siRNA Details
| Gene Symbol: CNIH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277197.1 | NP_001264126.1 | 3 | 343 | ||
| NM_001277198.1 | NP_001264127.1 | 3 | 343 | |||
| NM_001277199.1 | NP_001264128.1 | 3 | 621 | |||
| NM_001277200.1 | NP_001264129.1 | 3 | 343 | |||
| NM_014184.3 | NP_054903.1 | 3 | 343 | |||
| NR_102347.1 | 2 | 324 | ||||
| GenBank mRNA | AF161512.1 | - | 3 | 262 | ||
| AK024569.1 | - | 258 | ||||
| AK222634.1 | - | 3 | 248 | |||
| AK289973.1 | - | 3 | 277 | |||
| AK312064.1 | - | 3 | 268 | |||
| AL136930.1 | - | 3 | 283 | |||
| BC000573.1 | - | 3 | 258 | 3163724 | ||
| BI752106.1 | - | 3 | 259 | |||
| BQ051754.1 | - | 3 | 532 | |||
| BQ575976.1 | - | 3 | 559 | |||
| BX649076.1 | - | 3 | 247 | |||
| DA185135.1 | - | 3 | 344 | |||
| HY018269.1 | - | 2 | 240 | |||
| HY145928.1 | - | 3 | 268 | |||

CNIH4
Genomic Map