siRNA Details
| Gene Symbol: TMEM208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318217.1 | NP_001305146.1 | 4 | 487 | ||
| NM_014187.3 | NP_054906.2 | 4 | 370 | |||
| NR_134524.1 | 4 | 370 | ||||
| GenBank mRNA | BC003080.2 | - | 4 | 343 | 3050277 | |
| BC013412.2 | - | 4 | 322 | 3030307 | ||
| BC021109.1 | - | 4 | 326 | 3660106 | ||
| BF316569.1 | - | 4 | 296 | |||
| BG825054.1 | - | 4 | 448 | |||
| BI833541.1 | - | 4 | 329 | |||

TMEM208
Genomic Map