siRNA Details
| Gene Symbol: TMEM208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318217.1 | NP_001305146.1 | 6 | 729 | ||
| NM_014187.3 | NP_054906.2 | 6 | 612 | |||
| NR_134524.1 | 6 | 638 | ||||
| GenBank mRNA | AF161519.1 | - | 6 | 634 | ||
| BC003080.2 | - | 6 | 585 | 3050277 | ||
| BC013412.2 | - | 6 | 564 | 3030307 | ||
| BC021109.1 | - | 6 | 594 | 3660106 | ||
| BG825054.1 | - | 6 | 689 | |||
| BI833541.1 | - | 6 | 597 | |||

TMEM208
Genomic Map