siRNA Details
Gene Symbol: DROSHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100412.1 | NP_001093882.1 | 24, 25 | 3372 | ||
NM_013235.4 | NP_037367.3 | 24, 25 | 3407 | |||
XM_005248291.3 | XP_005248348.1 | 25, 26 | 3483 | |||
XM_005248292.3 | XP_005248349.1 | 25, 26 | 3459 | |||
XM_005248293.3 | XP_005248350.1 | 24, 25 | 3390 | |||
XM_005248294.3 | XP_005248351.1 | 23, 24 | 3279 | |||
XM_011514033.2 | XP_011512335.1 | 25, 26 | 3483 | |||
XM_017009399.1 | XP_016864888.1 | 24, 25 | 3366 | |||
XM_017009400.1 | XP_016864889.1 | 24, 25 | 3348 | |||
XM_017009401.1 | XP_016864890.1 | 23, 24 | 3255 | |||
XM_017009402.1 | XP_016864891.1 | 13, 14 | 1498 | |||
GenBank mRNA | AF116910.1 | - | 9, 10 | 936 | ||
AK001121.1 | - | 13, 14 | 1242 | |||
AK026563.1 | - | 4, 5 | 294 | |||
AK308093.1 | - | 23, 24 | 3181 | |||
BC024261.1 | - | 12, 13 | 1168 | |||
BC041162.1 | - | 11, 12 | 1093 | |||
BC054003.1 | - | 23, 24 | 3184 | 6148714 | ||
BX647724.1 | - | 24, 25 | 3275 |
