siRNA Details
| Gene Symbol: ANKRD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256182.1 | NP_001243111.1 | 10 | 1621 | ||
| NM_001256183.1 | NP_001243112.1 | 9 | 1547 | |||
| NM_013275.5 | NP_037407.4 | 9 | 1550 | |||
| XM_006721181.1 | XP_006721244.1 | 9 | 1327 | |||
| XM_006721184.2 | XP_006721247.1 | 8 | 1365 | |||
| XM_011523051.2 | XP_011521353.1 | 10 | 1610 | |||
| XM_011523053.2 | XP_011521355.1 | 10 | 1716 | |||
| XM_011523054.2 | XP_011521356.1 | 10 | 1630 | |||
| XM_011523055.2 | XP_011521357.1 | 11 | 1915 | |||
| XM_011523056.2 | XP_011521358.1 | 10 | 1652 | |||
| XM_011523057.2 | XP_011521359.1 | 9 | 1533 | |||
| XM_017023182.1 | XP_016878671.1 | 9 | 1983 | |||
| XM_017023183.1 | XP_016878672.1 | 10 | 1719 | |||
| XM_017023184.1 | XP_016878673.1 | 10 | 1716 | |||
| XM_017023185.1 | XP_016878674.1 | 8 | 1357 | |||
| XM_017023186.1 | XP_016878675.1 | 12 | 2093 | |||
| XM_017023187.1 | XP_016878676.1 | 8 | 1445 | |||
| GenBank mRNA | AB209186.1 | - | 8 | 1195 | ||
| AF121775.1 | - | 9 | 1306 | |||
| AK025964.1 | - | 6 | 918 | |||
| AK290994.1 | - | 8 | 1229 | |||
| AY373756.1 | - | 9 | 1553 | |||
| AY533563.1 | - | 10 | 1378 | |||
| BC017437.1 | - | 9 | 1519 | |||
| BC052290.1 | - | 9 | 1465 | |||
| BC069013.1 | - | 9 | 1465 | |||

ANKRD11
Genomic Map