siRNA Details
Gene Symbol: GRM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000842.4 | NP_000833.1 | 7 | 1978 | ||
NM_001143831.2 | NP_001137303.1 | 6 | 1780 | |||
XM_006718828.3 | XP_006718891.1 | 7 | 1976 | |||
XM_011542792.1 | XP_011541094.1 | 7 | 1978 | |||
XM_017017627.1 | XP_016873116.1 | 7 | 1976 | |||
GenBank mRNA | AK291362.1 | - | 7 | 1948 | ||
AY608336.1 | - | 6 | 1580 | |||
D28538.1 | - | 6 | 1730 | |||
D28539.1 | - | 6 | 1730 | |||
FJ588505.1 | - | 6 | 1580 | |||
FJ588506.1 | - | 6 | 1580 | |||
FJ588507.1 | - | 6 | 1580 | |||
FJ668670.1 | - | 6 | 1580 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
