siRNA Details
| Gene Symbol: GSTM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000848.3 | NP_000839.1 | 5 | 378 | ||
| NM_001142368.1 | NP_001135840.1 | 5 | 378 | |||
| GenBank mRNA | AK299482.1 | - | 5 | 507 | ||
| BC105038.1 | - | 5 | 309 | 8144041 | ||
| BC105066.1 | - | 5 | 309 | 8144069 | ||
| BC110380.1 | - | 5 | 331 | 5760414 | ||
| BM924181.1 | - | 5 | 344 | |||
| BT019947.1 | - | 5 | 284 | |||
| DA528798.1 | - | 5 | 378 | |||
| M63509.1 | - | 5 | 300 | |||

GSTM2
Genomic Map