siRNA Details
| Gene Symbol: TMOD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014547.4 | NP_055362.1 | 7 | 971 | ||
| XM_017022087.1 | XP_016877576.1 | 8 | 1002 | |||
| XM_017022088.1 | XP_016877577.1 | 7 | 843 | |||
| GenBank mRNA | AF177171.1 | - | 7 | 933 | ||
| AF237631.1 | - | 6 | 753 | |||
| AK310344.1 | - | 5 | 650 | |||
| AK312569.1 | - | 7 | 968 | |||
| BC020542.2 | - | 7 | 907 | 4421113 | ||
| BX647277.1 | - | 7 | 933 | |||

TMOD3
Genomic Map