siRNA Details
| Gene Symbol: CCDC106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013301.2 | NP_037433.2 | 6 | 1311 | ||
| XM_005258827.2 | XP_005258884.1 | 6 | 909 | |||
| XM_005258828.2 | XP_005258885.1 | 6 | 930 | |||
| GenBank mRNA | AF054984.1 | - | 6 | 864 | ||
| AK097095.1 | - | 5 | 1481 | |||
| BC000412.2 | - | 5 | 889 | 2961290 | ||
| BC008956.2 | - | 5 | 889 | 2961290 | ||
| GU354321.1 | - | 6 | 666 | |||
| GU354322.1 | - | 6 | 722 | |||
| GU354323.1 | - | 6 | 910 | |||
| U79303.1 | - | 6 | 864 | |||

CCDC106
Genomic Map